Gene Transcripts

Gene Symbol
Zkscan4
Entrez Gene ID
544922
Gene Name
zinc finger with KRAB and SCAN domains 4
Gene Aliases
AI841875, Gm11272, OTTMUSG00000000421
Chromosome Location
Chr. 13: 21478849 - 21485507 on Build GRCm38
UniGene ID
Mm.441300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039115.2 NP_001034204.1 2 786
GenBank mRNA BC141222.1 - 2 661
BC145543.1 - 2 574
BU148250.1 - 2 728

View Full Product Details