Gene Transcripts

Gene Symbol
Ptprh
Entrez Gene ID
545902
Gene Name
protein tyrosine phosphatase, receptor type, H
Gene Aliases
sap-1
Chromosome Location
Chr. 7: 4548614 - 4604041 on Build GRCm38
UniGene ID
Mm.188925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207270.2 NP_997153.2 20 3191
GenBank mRNA AB217856.1 - 19 2864
AK078884.1 - 19 2868

View Full Product Details