Gene Transcripts

Gene Symbol
Vmn2r75
Entrez Gene ID
546981
Gene Name
vomeronasal 2, receptor 75
Gene Aliases
EG546981
Chromosome Location
Chr. 7: 86148042 - 86171724 on Build GRCm38
UniGene ID
Mm.469835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102578.1 NP_001096048.1 3 751

View Full Product Details