Gene Transcripts

Gene Symbol
Pop4
Entrez Gene ID
66161
Gene Name
processing of precursor 4, ribonuclease P/MRP family, (S. cerevisiae)
Gene Aliases
1110023P21Rik, Rpp29
Chromosome Location
Chr. 7: 38262815 - 38271367 on Build GRCm38
UniGene ID
Mm.22284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025390.4 NP_079666.1 7 802
GenBank mRNA AK002855.1 - 7 803
AK003293.2 - 7 803
AK003908.1 - 7 816
AK054466.1 - 835
AK150459.1 - 7 809
AK152948.1 - 7 809
BC011465.1 - 7 818 3966371

View Full Product Details