Gene Transcripts

Gene Symbol
Mrpl4
Entrez Gene ID
66163
Gene Name
mitochondrial ribosomal protein L4
Gene Aliases
1110017G11Rik
Chromosome Location
Chr. 9: 21002605 - 21008839 on Build GRCm38
UniGene ID
Mm.427576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023167.2 NP_075656.2 1, 2 268
GenBank mRNA AB041599.1 - 1 19
AB049634.1 - 1, 2 267
AK002464.1 - 1, 2 104
AK003746.2 - 1, 2 226
BC023078.1 - 1, 2 49
BC037064.1 - 1, 2 101
BC039983.1 - 1, 2 236 4952757
BC051685.1 - 1, 2 81
BC061095.1 - 1, 2 84 6588836

View Full Product Details