Gene Transcripts

Gene Symbol
Prr3
Entrez Gene ID
75210
Gene Name
proline-rich polypeptide 3
Gene Aliases
4930540G07Rik, Cat56
Chromosome Location
Chr. 17: 35972539 - 35979825 on Build GRCm38
UniGene ID
Mm.40036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282012.1 NP_001268941.1 3 484
NM_001282013.1 NP_001268942.1 5 489
NM_001282024.1 NP_001268953.1 4 583
NM_145487.3 NP_663462.2 3 512
GenBank mRNA AJ504719.1 - 3 1281
AK132945.1 - 3 212
AK203788.1 - 1 37
BC016636.1 - 3 433 2650633
BU148736.1 - 5 446

View Full Product Details