Product Details

SNP ID
rs13481509
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:67216377 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTCAATGTTAATTGTATGGAGTATG[A/G]GTTTCTGGTTAAGTTTTCTAGCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mdga2 PubMed Links

Gene Details

Gene
Mdga2
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193266.1 Intron NP_001180195.2
NM_207010.2 Intron NP_996893.3

View Full Product Details