Product Details

SNP ID
rs6349956
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:24871639 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATCCAGGTTCTGCTTCATGAAGGC[A/G]GGCTTTGGCATGTCAGACATCTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Prkag2 PubMed Links

Gene Details

Gene
Prkag2
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170555.1 1763 Silent Mutation NP_001164026.1
NM_001170556.1 1763 Silent Mutation NP_001164027.1
NM_145401.2 1763 Silent Mutation NP_663376.2

View Full Product Details