Product Details

SNP ID
rs13478087
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:156133117 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGAGTCCTCAAAGTCTCTAAG[A/G]ACTACAGGCTTCTAAAGGCTCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
9430015G10Rik PubMed Links

Gene Details

Gene
9430015G10Rik
Gene Name
RIKEN cDNA 9430015G10 gene
There are no transcripts associated with this gene.

Gene
Rnf223
Gene Name
ring finger 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220499.1 948 UTR 3 NP_001207428.1

View Full Product Details