Product Details
- SNP ID
-
rs13476292
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:187919330 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGTGGTCTAGTTATCCAGGGCTC[A/G]GAACCTTCGCTAACTGCTGGACTAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Esrrg
PubMed Links
Gene Details
- Gene
- Esrrg
- Gene Name
- estrogen-related receptor gamma
View Full Product Details