Product Details

SNP ID
rs13475847
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:45912375 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGACACCAAATTCCAACCGGAAAT[A/G]AAACCACAGCCAATGACTGGAGAAC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
Slc40a1 PubMed Links

Gene Details

Gene
Slc40a1
Gene Name
solute carrier family 40 (iron-regulated transporter), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016917.2 952 Silent Mutation NP_058613.2

View Full Product Details