Product Details

SNP ID
rs6403469
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:11707169 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTAATTACAGCTGTAGAAATGGAGG[A/G]AGGATTGTTCAGTGTCTGTCCCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gem PubMed Links

Gene Details

Gene
Gem
Gene Name
GTP binding protein (gene overexpressed in skeletal muscle)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010276.4 Intron NP_034406.2

View Full Product Details