Product Details

SNP ID
rs13477553
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:9463709 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCATTTTCCACTTGCTGGTAGTATG[C/T]TTATCTACCAGTTCAAGTTTATTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Asph PubMed Links

Gene Details

Gene
Asph
Gene Name
aspartate-beta-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177849.1 Intron NP_001171320.1
NM_001177850.1 Intron NP_001171321.1
NM_001177851.1 Intron NP_001171322.1
NM_001177852.1 Intron NP_001171323.1
NM_001177853.1 Intron NP_001171324.1
NM_001177854.1 Intron NP_001171325.1
NM_001177855.1 Intron NP_001171326.1
NM_001177856.1 Intron NP_001171327.1
NM_023066.3 Intron NP_075553.2
NM_133723.2 Intron NP_598484.2

View Full Product Details