Product Details

SNP ID
rs13476444
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:38526867 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTAGATTGATTGAGATTGCCTGATA[C/T]ACCCAGCTGAAGTAGCTTTGGAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gm35808 PubMed Links

Gene Details

Gene
Gm35808
Gene Name
predicted gene, 35808
There are no transcripts associated with this gene.

Gene
Nek6
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159631.1 Intron NP_001153103.1
NM_021606.3 Intron NP_067619.1

View Full Product Details