Product Details

SNP ID
rs4222719
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:156327389 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCAGAATTTTTCATACCAAGT[C/T]CTAATTGCTTGCTGTCATGTACACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Axdnd1 PubMed Links

Gene Details

Gene
Axdnd1
Gene Name
axonemal dynein light chain domain containing 1
There are no transcripts associated with this gene.

Gene
Nphs2
Gene Name
nephrosis 2, podocin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130456.3 2462 UTR 3 NP_569723.1

View Full Product Details