Product Details
- SNP ID
-
rs4163058
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:13269665 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTATTCAGGGCATACCACTGTCT[G/A]CATCTGTTTGACTGGAGAAGAATAT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mkl2
PubMed Links
Gene Details
- Gene
- Mkl2
- Gene Name
- MKL/myocardin-like 2
View Full Product Details