Product Details

SNP ID
rs13482164
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:46847514 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GATTTGGGGCTCATCTCTGTCCCCA[A/G]GATAGCTCATTGTATATATGCCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cgrrf1 PubMed Links

Gene Details

Gene
Cgrrf1
Gene Name
cell growth regulator with ring finger domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026832.3 Intron NP_081108.2

View Full Product Details