Product Details
- SNP ID
-
rs3672124
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110615073 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCCACTGTCAGTTTTGACACTGA[T/C]ACAGGTAACCCAGAACCAATAGCCA
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dync1h1
PubMed Links
Gene Details
- Gene
- Dync1h1
- Gene Name
- dynein cytoplasmic 1 heavy chain 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030238.2 |
1161 |
Silent Mutation |
|
|
NP_084514.2 |
View Full Product Details