Product Details

SNP ID
rs3688141
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:75192214 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGATGACACCGATTCAGACAGTAAG[A/G]TGATCTTGCCCTTTATAAAACTCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ltbp1 PubMed Links

Gene Details

Gene
Ltbp1
Gene Name
latent transforming growth factor beta binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019919.3 Intron NP_064303.2
NM_206958.2 Intron NP_996841.1

View Full Product Details