Product Details

SNP ID
rs13476485
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:49535259 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGCACTGTTTTACCCAGGTGC[C/T]GTCTCCAGCTTCAGAGGCTGAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Epc2 PubMed Links

Gene Details

Gene
Epc2
Gene Name
enhancer of polycomb homolog 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172663.4 1465 Missense Mutation NP_766251.3

View Full Product Details