Product Details
- SNP ID
-
rs4171545
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:33698177 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCAGGGGCACCAGCTCTGCTTC[G/T]CTGAGCTTCTCACACCCCAGAGGGA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
Heg1
PubMed Links
Gene Details
- Gene
- Heg1
- Gene Name
- HEG homolog 1 (zebrafish)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175256.5 |
|
Intron |
|
|
NP_780465.4 |
View Full Product Details