Product Details

SNP ID
rs13481369
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:29570417 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGGGCTTTGTATTATCAGCAC[A/G]AGCAACTACAATACTGAAGTCAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Myt1l PubMed Links

Gene Details

Gene
Myt1l
Gene Name
myelin transcription factor 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093775.1 Intron NP_001087244.1
NM_001093776.1 Intron NP_001087245.1
NM_001093778.1 Intron NP_001087247.1
NM_008666.3 Intron NP_032692.2

View Full Product Details