Product Details

SNP ID
rs3706453
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:9999862 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTTGACTACCACATGTTCAAGTCT[G/T]CACTCACGCACAAGAATGCATGCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Ppp1r42 PubMed Links

Gene Details

Gene
Ppp1r42
Gene Name
protein phosphatase 1, regulatory subunit 42
There are no transcripts associated with this gene.

View Full Product Details