Product Details

SNP ID
rs3714096
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:35696573 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTATGGAGCTTATAACCCAGCCT[G/A]GTAATGCAGATGCGGTGGTGTTTGA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Dnajc18 PubMed Links

Gene Details

Gene
Dnajc18
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029669.4 Intron NP_083945.1

View Full Product Details