Product Details

SNP ID
rs4222872
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:179686870 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTATGATTTTATTGCTTGTGA[A/G]AGGAAACTAACATTCTCCTTACGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gm1305 PubMed Links

Gene Details

Gene
Gm1305
Gene Name
predicted gene 1305
There are no transcripts associated with this gene.

Gene
Sccpdh
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178653.3 1608 UTR 3 NP_848768.1

View Full Product Details