Product Details
- SNP ID
-
rs3023068
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52232248 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGACTTGACACTGAGAAAGATCT[C/T]AACCTCTATAGAATTTTAGCGATAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Hoxa10
PubMed Links
Gene Details
- Gene
- Hoxa10
- Gene Name
- homeobox A10
- Gene
- Hoxa11
- Gene Name
- homeobox A11
There are no transcripts associated with this gene.
- Gene
- Hoxa9
- Gene Name
- homeobox A9
There are no transcripts associated with this gene.
- Gene
- Mir196b
- Gene Name
- microRNA 196b
There are no transcripts associated with this gene.
View Full Product Details