Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
102127264
Gene Name
stem-loop binding protein
Gene Aliases
-
Chromosome Location
Chr.5: 1701841 - 1727540 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015449988.1 XP_015305474.1 4 - 5 418 115

View Full Product Details