Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR2
Entrez Gene ID
102146068
Gene Name
toll like receptor 2
Gene Aliases
-
Chromosome Location
Chr.5: 152925504 - 152947212 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319613.1 NP_001306542.1 1 - 1 1964 114
XM_005556107.2 XP_005556164.1 3 - 3 2197 114
XM_005556108.2 XP_005556165.1 2 - 2 2048 114
GenBank mRNA AB445629.1 - 1 - 1 1964 114
AY045573.1 - 3 - 3 2127 114

View Full Product Details