Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAMPT
Entrez Gene ID
102144763
Gene Name
nicotinamide phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.3: 138700007 - 138738545 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550471.2 XP_005550528.1 9 - 10 1533 109
XM_015447907.1 XP_015303393.1 10 - 11 2009 109

View Full Product Details