Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYVE1
Entrez Gene ID
102136766
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
-
Chromosome Location
Chr.14: 57230042 - 57246498 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578624.2 XP_005578681.1 5 - 6 1070 111

View Full Product Details