Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF77
Entrez Gene ID
102120956
Gene Name
zinc finger protein 77
Gene Aliases
-
Chromosome Location
Chr.19: 2885935 - 2900358 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587487.2 XP_005587544.1 2 - 3 410 75

View Full Product Details