Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS10
Entrez Gene ID
102133236
Gene Name
regulator of G protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.9: 118738696 - 118781349 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566569.2 XP_005566626.1 1 - 2 133 64

View Full Product Details