Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
102115995
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 9144407 - 9160670 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591219.2 XP_005591276.1 1 - 2 106 94

View Full Product Details