Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MID2
Entrez Gene ID
102122299
Gene Name
midline 2
Gene Aliases
-
Chromosome Location
Chr.X: 104926324 - 105027481 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594318.2 XP_005594375.1 9 - 10 1905 63
XM_005594319.2 XP_005594376.1 9 - 10 1815 63
XM_005594320.2 XP_005594377.1 9 - 10 1905 63

View Full Product Details