Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXK2
Entrez Gene ID
102130025
Gene Name
forkhead box K2
Gene Aliases
-
Chromosome Location
Chr.16: 80210346 - 80290534 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437983.1 XP_015293469.1 1 - 2 355 82

View Full Product Details