Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLA2G2E
Entrez Gene ID
102134757
Gene Name
phospholipase A2 group IIE
Gene Aliases
-
Chromosome Location
Chr.1: 208505331 - 208510558 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544568.2 XP_005544625.1 2 - 3 1601 149

View Full Product Details