Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE2
Entrez Gene ID
102115013
Gene Name
jade family PHD finger 2
Gene Aliases
PHF15
Chromosome Location
Chr.6: 133578940 - 133637170 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557789.2 XP_005557846.2 2 - 3 304 79
XM_005557790.2 XP_005557847.1 2 - 3 305 79
XM_005557791.2 XP_005557848.1 2 - 3 306 79
XM_005557792.2 XP_005557849.1 2 - 3 306 79
XM_005557795.2 XP_005557852.1 2 - 3 409 79
XM_005557796.2 XP_005557853.1 2 - 3 170 79
XM_005557798.2 XP_005557855.1 2 - 3 171 79
XM_005557799.2 XP_005557856.1 2 - 3 180 79
XM_015451895.1 XP_015307381.1 3 - 4 409 79
XM_015451896.1 XP_015307382.1 2 - 3 409 79

View Full Product Details