Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIAO2B
Entrez Gene ID
102119717
Gene Name
cytosolic iron-sulfur assembly component 2B
Gene Aliases
FAM96B
Chromosome Location
Chr.20: 55072461 - 55075282 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592188.2 XP_005592245.1 3 - 4 868 97

View Full Product Details