Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX4
Entrez Gene ID
102122465
Gene Name
T-box transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.16: 46342269 - 46372619 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438386.1 XP_015293872.1 5 - 6 2622 76
XM_015438388.1 XP_015293874.1 5 - 6 2621 76

View Full Product Details