Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
102133655
Gene Name
vesicle amine transport 1 like
Gene Aliases
-
Chromosome Location
Chr.20: 66049785 - 66243231 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592608.2 XP_005592665.1 4 - 5 865 85

View Full Product Details