Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH3
Entrez Gene ID
102133089
Gene Name
G-patch domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 201378444 - 201391179 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544305.2 XP_005544362.1 4 - 5 1873 128
GenBank mRNA AB169881.1 - 4 - 5 1151 128

View Full Product Details