Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OGFRL1
Entrez Gene ID
102115228
Gene Name
opioid growth factor receptor like 1
Gene Aliases
-
Chromosome Location
Chr.4: 102625838 - 102646230 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552588.2 XP_005552645.1 1 - 2 839 67
XM_005552589.2 XP_005552646.1 1 - 2 839 67

View Full Product Details