Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PODNL1
Entrez Gene ID
102136838
Gene Name
podocan like 1
Gene Aliases
-
Chromosome Location
Chr.19: 14268656 - 14275370 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440702.1 XP_015296188.1 6 - 7 871 90
XM_015440703.1 XP_015296189.1 6 - 7 871 90

View Full Product Details