Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM9A
Entrez Gene ID
102123788
Gene Name
family with sequence similarity 9 member A
Gene Aliases
-
Chromosome Location
Chr.X: 6429116 - 6438513 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592929.1 XP_005592986.1 4 - 5 452 160

View Full Product Details