Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H19orf47
Entrez Gene ID
102134434
Gene Name
chromosome 19 open reading frame, human C19orf47
Gene Aliases
-
Chromosome Location
Chr.19: 40798189 - 40827169 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589259.2 XP_005589316.1 2 - 3 206 108

View Full Product Details