Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHISA6
Entrez Gene ID
102134511
Gene Name
shisa family member 6
Gene Aliases
-
Chromosome Location
Chr.16: 11406719 - 11736676 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583173.2 XP_005583230.2 1 - 2 392 89

View Full Product Details