Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
B3GALNT1
Entrez Gene ID
102122721
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases
-
Chromosome Location
Chr.2: 63478332 - 63502736 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005546263.2 XP_005546320.1 1 - 1 105 108

View Full Product Details