Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TARM1
Entrez Gene ID
102122004
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Gene Aliases
-
Chromosome Location
Chr.19: 54917348 - 54932938 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590275.2 XP_005590332.1 2 - 3 181 64

View Full Product Details