Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD34B
Entrez Gene ID
102134724
Gene Name
ankyrin repeat domain 34B
Gene Aliases
-
Chromosome Location
Chr.6: 78831763 - 78839544 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015451643.1 XP_015307129.1 2 - 2 2013 93

View Full Product Details