Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
102119602
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C12H2orf62
Chromosome Location
Chr.12: 108007787 - 108021852 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287667.1 NP_001274596.1 2 - 3 212 64
XM_005574280.2 XP_005574337.1 3 - 4 428 64
GenBank mRNA AB071068.1 - 2 - 3 212 64

View Full Product Details